Anti-GAB2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Proveedor: Biorbyt
BIRBORB8915-100EA 0 EUR
BIRBORB8915-100
Anti-GAB2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: GAB2 (GRB2-associated binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat