Anti-APOB Mouse Monoclonal Antibody [clone: 1D1]

Proveedor: Biorbyt
BIRBORB8250-100EA 667 EUR
BIRBORB8250-100
Anti-APOB Mouse Monoclonal Antibody [clone: 1D1]
Anticuerpos
Western Blot: 1:10001:3000

Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: monoclonal
Clone: 1D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Murine)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR