Anti-HBsAg Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB8243-100EA 667 EUR
BIRBORB8243-100
Anti-HBsAg Mouse monoclonal antibody
Anticuerpos
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: HBsAg
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR