Anti-CD27 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB8169-100EA 0 EUR
BIRBORB8169-100
Anti-CD27 Mouse Monoclonal Antibody
Anticuerpos
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: CD27 (CD27 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR