Anti-MS4A1 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Proveedor: Biorbyt
BIRBORB8154-100EA 667 EUR
BIRBORB8154-100
Anti-MS4A1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR