Anti-CD7 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB8117-100EA 550 EUR
BIRBORB8117-100
Anti-CD7 Mouse Monoclonal Antibody
Anticuerpos
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: CD7 (CD7 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR