Anti-APBB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
BIRBORB8082-100EA 385 EUR
BIRBORB8082-100
Anti-APBB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: APBB1 (amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR