Anti-S100P Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB6915-100EA 0 EUR
BIRBORB6915-100 BIRBORB6915-200
Anti-S100P Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500
Type: Primary
Antigen: S100P (S100 calcium binding protein P)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human