Anti-RIPK2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB6880-100EA 334 EUR
BIRBORB6880-100
Anti-RIPK2 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500

Type: Primary
Antigen: RIPK2 (receptor-interacting serine-threonine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR