Anti-PXN Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB6652-100EA 232 EUR
BIRBORB6652-100 BIRBORB6652-200
Anti-PXN Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PXN (paxillin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR