Anti-P2RX3 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB6566-5EA 51 EUR
BIRBORB6566-5 BIRBORB6566-20 BIRBORB6566-200 BIRBORB6566-100
Anti-P2RX3 Rabbit Polyclonal Antibody
Anticuerpos
Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: P2RX3 (purinergic receptor P2X, ligand-gated ion channel, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR