Anti-RELA Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB6502-100EA 0 EUR
BIRBORB6502-100
Anti-RELA Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR