Anti-MAPKAPK2 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB6385-200EA 0 EUR
BIRBORB6385-200 BIRBORB6385-100
Anti-MAPKAPK2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MAPKAPK2 (mitogen-activated protein kinase-activated protein kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat