Anti-LYN Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB6339-100EA 0 EUR
BIRBORB6339-100
Anti-LYN Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR