Anti-LYN Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB6338-5EA 51 EUR
BIRBORB6338-5 BIRBORB6338-200 BIRBORB6338-20 BIRBORB6338-100
Anti-LYN Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR