Anti-STK11 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB6322-100EA 0 EUR
BIRBORB6322-100
Anti-STK11 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: STK11 (serine/threonine kinase 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR