Anti-GJA4 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB6190-100EA 312 EUR
BIRBORB6190-100 BIRBORB6190-200
Anti-GJA4 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: GJA4 (gap junction protein, alpha 4, 37kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR