Anti-EIF2AK4 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB6077-100EA 312 EUR
BIRBORB6077-100
Anti-EIF2AK4 Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: EIF2AK4 (eukaryotic translation initiation factor 2 alpha kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR