Anti-EIF2A Rabbit polyclonal antibody
Proveedor: Biorbyt
BIRBORB5997-100EA 0 EUR
BIRBORB5997-100 BIRBORB5997-200
Anti-EIF2A Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: EIF2A (eukaryotic translation initiation factor 2A, 65kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now