Anti-PRKDC Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB5979-100EA 305 EUR
BIRBORB5979-100
Anti-PRKDC Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:1000

Type: Primary
Antigen: PRKDC (protein kinase, DNA-activated, catalytic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR