Anti-CLDN4 Rabbit polyclonal antibody
Proveedor: Biorbyt
BIRBORB5879-100EA 0 EUR
BIRBORB5879-100 BIRBORB5879-200
Anti-CLDN4 Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CLDN4 (claudin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now