Anti-CDC25A Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB5827-100EA 0 EUR
BIRBORB5827-100 BIRBORB5827-200
Anti-CDC25A Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now