Anti-CD19 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB5789-100EA 306 EUR
BIRBORB5789-100
Anti-CD19 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:2000

Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR