Anti-Kappa light chain Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB5667-200EA 0 EUR
BIRBORB5667-200 BIRBORB5667-100
Anti-Kappa light chain Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: Kappa Light Chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human