Anti-IKBKE Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB5531-100EA 0 EUR
BIRBORB5531-100 BIRBORB5531-200
Anti-IKBKE Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: IKBKE (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat