Anti-HSD11B1 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB5472-200EA 0 EUR
BIRBORB5472-200 BIRBORB5472-100
Anti-HSD11B1 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: HSD11B1 (hydroxysteroid (11-beta) dehydrogenase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat