Anti-rHPV Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB5461-200EA 508 EUR
BIRBORB5461-200 BIRBORB5461-100
Anti-rHPV Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500

Type: Primary
Antigen: rHPV
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR