Anti-RIT1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
BIRBORB5255-100EA 385 EUR
BIRBORB5255-100
Anti-RIT1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: RIT1 (Ras-like without CAAX 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR