Anti-FBP1 Rabbit polyclonal antibody
Proveedor: Biorbyt
BIRBORB5232-200EA 0 EUR
BIRBORB5232-200 BIRBORB5232-100
Anti-FBP1 Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: FBP1 (fructose-1,6-bisphosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat