Anti-NFX1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Proveedor: Biorbyt
BIRBORB5144-100EA 0 EUR
BIRBORB5144-100
Anti-NFX1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: NFX1 (nuclear transcription factor, X-box binding 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human