Anti-CCDC30 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: Biorbyt
BIRBORB5056-100EA 385 EUR
BIRBORB5056-100
Anti-CCDC30 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CCDC30 (coiled-coil domain containing 30)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR