Anti-SSH2 Mouse Monoclonal Antibody [clone: 1E11]

Proveedor: Abnova
ABNOH00085464-M07EA 394 EUR
ABNOH00085464-M07
Anti-SSH2 Mouse Monoclonal Antibody [clone: 1E11]
Anticuerpos
Type: Primary
Antigen: SSH2
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR