Anti-SUFU Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOH00051684-D01PEA 380 EUR
ABNOH00051684-D01P
Anti-SUFU Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SUFU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR