Anti-KLK4 Mouse Monoclonal Antibody [clone: 2A4]

Proveedor: Abnova
ABNOH00009622-M09EA 380 EUR
ABNOH00009622-M09
Anti-KLK4 Mouse Monoclonal Antibody [clone: 2A4]
Anticuerpos
Type: Primary
Antigen: KLK4
Clonality: Monoclonal
Clone: 2A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR