Anti-PAPPA Mouse Monoclonal Antibody [clone: 1G3]

Proveedor: Abnova
ABNOH00005069-M01EA 380 EUR
ABNOH00005069-M01
Anti-PAPPA Mouse Monoclonal Antibody [clone: 1G3]
Anticuerpos
Type: Primary
Antigen: PAPPA
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR