Anti-OCLN Mouse Polyclonal Antibody

Proveedor: Abnova
ABNOH00004950-A01EA 250 EUR
ABNOH00004950-A01
Anti-OCLN Mouse Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: OCLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR