Anti-CTSG Rabbit Polyclonal Antibody
Proveedor: Abnova
ABNOH00001511-D01PEA 380 EUR
ABNOH00001511-D01P
Anti-CTSG Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CTSG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse