Anti-RYBP Mouse Monoclonal Antibody [clone: OTI5A9]

Proveedor: OriGene
ORIGTA504654EA 603 EUR
ORIGTA504654
Anti-RYBP Mouse Monoclonal Antibody [clone: OTI5A9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RYBP (RING1 and YY1 binding protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR