Anti-CPA2 Mouse Monoclonal Antibody [clone: OTI1D8]

Proveedor: OriGene
TA504645
ORIGTA504645EA 593 EUR
ORIGTA504645
Anti-CPA2 Mouse Monoclonal Antibody [clone: OTI1D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CPA2 (carboxypeptidase A2 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR