Anti-SAT2 Mouse Monoclonal Antibody [clone: OTI2B3]

Proveedor: OriGene
TA504632
ORIGTA504632EA 598 EUR
ORIGTA504632
Anti-SAT2 Mouse Monoclonal Antibody [clone: OTI2B3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: SAT2 (spermidine/spermine N1-acetyltransferase family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR