Anti-MIEN1 Mouse Monoclonal Antibody [clone: OTI1G2]

Proveedor: OriGene
ORIGTA504614EA 603 EUR
ORIGTA504614
Anti-MIEN1 Mouse Monoclonal Antibody [clone: OTI1G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MIEN1 (migration and invasion enhancer 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR