Anti-SPINT1 Mouse Monoclonal Antibody [clone: OTI4E1]

Proveedor: OriGene
ORIGTA504605EA 603 EUR
ORIGTA504605
Anti-SPINT1 Mouse Monoclonal Antibody [clone: OTI4E1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: SPINT1 (serine peptidase inhibitor, Kunitz type 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR