Anti-SERPINE2 Mouse Monoclonal Antibody [clone: OTI1G8]

Proveedor: OriGene
TA504601
ORIGTA504601EA 616 EUR
ORIGTA504601
Anti-SERPINE2 Mouse Monoclonal Antibody [clone: OTI1G8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SERPINE2 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR