Anti-HSD17B2 Mouse Monoclonal Antibody [clone: OTI4F10]

Proveedor: OriGene
ORIGTA504565EA 603 EUR
ORIGTA504565
Anti-HSD17B2 Mouse Monoclonal Antibody [clone: OTI4F10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HSD17B2 (hydroxysteroid (17-beta) dehydrogenase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR