Anti-CST2 Mouse Monoclonal Antibody [clone: OTI2D6]

Proveedor: OriGene
ORIGTA504560EA 586 EUR
ORIGTA504560
Anti-CST2 Mouse Monoclonal Antibody [clone: OTI2D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: CST2 (cystatin SA)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR