Anti-DNAAF1 Mouse Monoclonal Antibody [clone: OTI4E4]
Proveedor: OriGene
ORIGTA504535EA 603 EUR
ORIGTA504535
Anti-DNAAF1 Mouse Monoclonal Antibody [clone: OTI4E4]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: DNAAF1 (dynein, axonemal, assembly factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human