Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI2E3]

Proveedor: OriGene
ORIGTA504517EA 586 EUR
ORIGTA504517
Anti-CPA1 Mouse Monoclonal Antibody [clone: OTI2E3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CPA1 (carboxypeptidase A1 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR