Anti-SCHIP1 Mouse Monoclonal Antibody [clone: OTI1C4]

Proveedor: OriGene
ORIGTA504470EA 586 EUR
ORIGTA504470
Anti-SCHIP1 Mouse Monoclonal Antibody [clone: OTI1C4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SCHIP1 (schwannomin interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR