Anti-ACD Mouse Monoclonal Antibody [clone: OTI2B1]

Proveedor: OriGene
ORIGTA504379EA 608 EUR
ORIGTA504379
Anti-ACD Mouse Monoclonal Antibody [clone: OTI2B1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: ACD (adrenocortical dysplasia homolog (mouse))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR