Anti-PSMB9 Mouse Monoclonal Antibody [clone: OTI1G7]

Proveedor: OriGene
ORIGTA504350EA 603 EUR
ORIGTA504350
Anti-PSMB9 Mouse Monoclonal Antibody [clone: OTI1G7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PSMB9 (proteasome (prosome, macropain) subunit, beta type, 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR